Learn More
BLMH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14941-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The normal physiological role of BLM hydrolase is unknown, but it catalyzes the inactivation of the antitumor drug BLM (a glycopeptide) by hydrolyzing the carboxamide bond of its B-aminoalaninamide moiety thus protecting normal and maligt cells from BLM toxicity.Specifica
| BLMH | |
| Polyclonal | |
| Unconjugated | |
| BLMH | |
| BH, bleomycin hydrolase, BLM hydrolase, BLMH, BMH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 104184, 287552, 642 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P70645, Q13867, Q8R016 | |
| Blmh | |
| BLMH Fusion Protein Ag6888 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |