missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Blooms Syndrome Protein Blm Antibody (BFL 103) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals (Bio-Techne) NBP2-50122JF525
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Blooms Syndrome Protein Blm Monoclonal antibody specifically detects Blooms Syndrome Protein Blm in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifica
| Blooms Syndrome Protein Blm | |
| Monoclonal | |
| Janelia Fluor 525 | |
| Bloom syndrome, Bloom syndrome, RecQ helicase-like, BSBloom syndrome protein, DNA helicase, RecQ-like type 2, EC 3.6.1, EC 3.6.4.12, MGC126616, MGC131618, RecQ protein-like 3, RecQ2, RECQ2MGC131620, RECQL3RECQL2 | |
| Recombinant Full Length Bloom's Protein | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Genes Sensitive to DNA Damaging Agents, Phospho Specific, Stem Cell Markers, Tumor Suppressors | |
| 641 | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| BFL 103 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto