Learn More
Brady™ Kit stampante monocromatica per etichette BMP21
Descrizione
- Designed for Laboratory: additional laboratory symbols in font 9 and up, improved barcode quality, ability to preset custom label lengths
- Labeling: materials available for applications such as cryo vials and slides; Greek symbols commonly used in laboratory, intuitive menu structure for symbols
- Vial label function automatically cuts the tape according to the size of the vial; Time & Date stamp; Automated serialisation
- Laboratory specific materials designed to withstand harsh laboratory environments. High quality, long lasting labels that stick and stay stuck
- Prints durable true-sized 6 – 19 mm wide identification to maximise amount of data. Automatic label formatting for commonly used label types in laboratory
- Long life rechargeable Lithium-ion battery - accessories sold separately. Battery included in the Starter Kit only
- 3 line LCD display provides clear visibility
- 2 year Warranty
Specifica
Specifica
| Product Type | Kit per stampante per etichette |
| Tipo di display | Monocolor LCD |
| Da utilizzare con (apparecchiatura) | Vial and Tube Labels, Slide Labels, Straw Labels, Bottle and Dish Labels |
| Print Technology | Trasferimento termico |
| Larghezza (sistema metrico) | 125 mm |
| Altezza (sistema metrico) | 235 mm |
| Profondità (sistema metrico) | 50 mm |
| Dimensioni (L x P) | 235 x 125 mm |
| Dimensioni (L x P x A) | 50 x 125 x 235 mm |
| Capacità colore | Monocromatico |
| Vedi altri risultati |