missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BNIP3 Antibody (ANa40), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NB100-1543R
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
BNIP3 Monoclonal specifically detects BNIP3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifica
| BNIP3 | |
| Monoclonal | |
| DyLight 550 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| BNIP3 | |
| Recombinant fragment (Residues 1-163 of Human BNIP3). | |
| 0.1 mL | |
| Primary | |
| BNIP3 (ANa40) | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| ANa40 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| BCL2/adenovirus E1B 19 kDa protein-interacting protein 3, BCL2/adenovirus E1B 19kDa interacting protein 3, BCL2/adenovirus E1B 19kD-interacting protein 3, NIP3 | |
| Mouse | |
| Protein A or G purified | |
| Apoptosis, Autophagy, Cancer, Hypoxia, Phospho Specific, Tumor Suppressors | |
| 664.0 | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction