Learn More
Grant Instruments™ Test Tube Rack for Unstirred Water Baths
Click to view available options
Capacità:
105 0.5 mL Tubes
12 30 mm Tubes
18 25 mm Tubes
32 19 mm Tubes
36 16 mm Tubes
55 13 mm Tubes
65 1.5 mL Tubes
84 10 mm Tubes
Dimensione della confezione:
Pezzo
Descrizione
Keeps test tubes upright and secure in the bath.
- Compatible with a wide range of Grant Unstirred Water Baths
- Holds up to 105 test tubes depending on size
- Dual slotted trays keep tubes secure and prevent tipping
Specifica
Specifica
| Capacità | 105 0.5 mL Tubes |