Learn More
Bohlender™ BOLA™ Reactor Lids for Glass Reactors DN 60
Click to view available options
Dettagli porta:
NPT 1/2 in. aside the shaft
NPT 1/4 in. to shaft, NPT 1/2 aside
Dimensione della confezione:
Pezzo
Descrizione
- Suitable for glass reactors DN 60 with SCHOTT flat flange
- With screw-in threads for connection of transition fittings and a center neck NPT 1/2 in.
- Easy assembly
- Flexibly expandable
- Completely made of PTFE
- Universal chemical resistance
- With connectors for the use of existing equipment with ground joint or GL thread
Thread connections in detail:
- Centric thread connection (purple) for insertion of a stirrer bearing
- Vertical thread connection (blue) with parallel alignment to the stirrer shaft
- Inclined thread connection (yellow) with direction straight to the stirrer shaft
- Inclined thread connection (green) with direction aside the stirrer shaft
Specifica
Specifica
| Descrizione | Center Neck 1/2 in., Lateral Necks 3 x 1/2 in. |
| Da utilizzare con (apparecchiatura) | Glass Reactors DN 60 with SCHOTT Flat Flange |
| Product Type | 4 x Female Thread Center Neck |
| Materiale | PTFE |
| Dettagli porta | NPT 1/2 in. aside the shaft |