Learn More
Bohlender™ BOLA™ Sleeves with Gripping Ring
Click to view available options
N. giunto rastremato standard:
US 24/40
Dimensione della confezione:
pezzi 2
Descrizione
- Made of PTFE
- Gripping ring and sealing rings on the outside
- High-vacuum tight sealing
- Low leak rate: <1 x 0.0001 mbar x l x s-1
- Solid construction for continued use
- Sealing rings are easy accessible and therefore easy to clean
Specifica
Specifica
| Materiale | PTFE |
| N. giunto rastremato standard | US 24/40 |
| Tipo | Sleeve with Gripping Ring |