Learn More
Bohlender™ BOLA™ Spherical Ground Joint Sleeves
Click to view available options
N. giunto rastremato standard:
S 13
S 19
S 29
S 40
Dimensione della confezione:
pezzi 2
pezzi 3
Descrizione
- With gripping ring and sealing rings on the outside
- High-vacuum tight sealing
- Low leak rate: <1 x 0.01 mbar x l x s-1
- Solid construction for continued use
Specifica
Specifica
| Materiale | PTFE |
| Product Type | Ground Joint Sleeves |
| N. giunto rastremato standard | S 13 |