Learn More
Bohlender™ BOLA Standard Construction Kit
For making flanges at the ends of plastic tubing (ex. PTFE, PFA or FEP)
Marca: Bohlender™ F700-04
Dettagli aggiuntivi : Peso : 0.50000kg
Descrizione
Consisting of:
- 1x BOLA Thermoelectric Flanging Tool with exchangeable flanging tip for tubing I.D. 1.6mm and tubing holder for tubing O.D. 1.6mm (1/16 in.) and 3.2 mm (1/8 in.)
- 1 x BOLA Tube End Fitting Set
- 5 x BOLA Plugs
- 10 x BOLA Miniature Couplings (straight)
- 2 x Miniature Couplings (T)
- 1 x BOLA Miniature Coupling (cross)
- 10 metres of PTFE tubing (I.D. 1.6mm, O.D. 3.2mm)
Specifica
230V / 50 Hz | |
Not Autoclavable | |
For making flanges at the ends of plastic tubing |
BOLA Standard Construction Kit | |
Plastic tubing |