Learn More
Thermo Scientific™ Bonded AVCS Pre-slit Caps
Click to view available options
Colore:
Blue
Green
Red
Yellow
Dimensione della confezione:
pezzi 100
Descrizione
Significantly reduce the error rate of analysis that results from evaporation of solvent in vials which are not sealed correctly.
- Re-analysis costs are reduced, sample data integrity increases, and sample throughput increases.
- AVCS closures seal as well as crimp top vials.
- AVCS design allows for consistent sealing, eliminating user-to-user variance.
Specifica
Specifica
| Colore | Blue |
| Diametro (sistema metrico) esterno | 9 mm |
| Pre-intagliato | Yes |
| Setto | PTFE/Silicone |
| Product Line | Surestop |
| Quantità | 100/Pk. |