missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD™ Bordet Gengou Agar with 15% Sheep Blood, Plates
Click to view available options
:
Bordet Gengou Agar with 15% Sheep Blood: 20 x 90mm Plates
Description
All Bordetella species (e.g. B. pertussis and B. parapertussis) are respiratory pathogens, residing in ciliated epithelial cells of the respiratory tract. Bordet Gengou Agar Base is a modification of the medium originally described by Bordet and Gengou for the cultivation of Bordetella pertussis.
- Prepared according to the American Public Health Association formulation
- Infusion from potato provides nitrogen and vitamins
- Potato starch absorbs fatty acids that may inhibit growth of B. pertussis
- Glycerol is a carbon source
- Sodium chloride maintains the osmotic balance of the medium
- The addition of 15% sheep blood provides essential complex growth factors
- Cefalexin is a selective agent to partially suppress normal respiratory tract flora
Supplied in ready to use Stacker™ plates
- Prepared plated media are ready for immediate use
- Microbiologically controlled for uncompromised isolation of microorganisms
- Specialized lid and bottom allow plates to interlock and stack without slipping
- Specially engineered design easily separates without agitating the sample
Specifications
Specifications
| Certificazione/Conformità | APHA |
| Content And Storage | Store in dark in original sleeve in 2°-8° C |
| Formato | Prepared culture plates |
| Product Type | Agar |
| Target Organism Class | Bordetella species |
| Formulazione | Per 1 L prepared: Potato Infusion 4.5 g, Pancreatic Digest of Casein 5 g, Peptic Digest of Animal Tissue 5 g, Sodium Chloride 5.5 g, Agar 20 g, Cefalexin 0.04 g, Glycerol 10 mL, Sheep Blood (defibrinated) 15% |
| Da utilizzare con (applicazione) | Isolation of Bordetella pertussis and B. parapertussis from nasopharyngeal specimens |
| Condizioni di incubazione | 34°-36° C in ambient air in a moist chamber |
| Tempo di incubazione | 7-10 days |
| Tipo di imballaggio | 10 lidded plates per plastic sleeve |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction