Learn More
BORIS Mouse anti-Human, CoraLite 488, Clone: 2B12E2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL488-60079-100UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The BORIS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BORIS. This antibody recognizes human, mouse antigen. The BORIS antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifica
| BORIS | |
| Monoclonal | |
| 2000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse |
| Immunofluorescence | |
| 2B12E2 | |
| CoraLite 488 | |
| 100 μL | |
| 140690 | |
| IgG1 |