Learn More
IKA™ Borosilicate Glass Coated Receiving Flask
Shop All IKA Products
missing translation for 'orderingAttributeHoverText'
Volume (sistema metrico):
100 mL
250 mL
500 mL
3000 mL
missing translation for 'unitSize'
Pezzo
Specifications
Specifications
| Materiale | Borosilicate Glass |
| Tipo | Receiving Flask |
| Quantità | 1 Each |
| Volume (sistema metrico) | 500 mL |