Learn More
Cytiva Borosilicate Glass Coated Tricorn™ Glass Tube with Protective Plastic Film
Codice prodotto. 10388649 Sfoglia Tutto Cytiva Prodotti
Click to view available options
Lunghezza (sistema metrico):
20 mm
50 mm
100 mm
150 mm
200 mm
300 mm
600 mm
Diametro (sistema metrico):
5 mm
10 mm
Dimensione della confezione:
Pezzo
Codice prodotto. 10388649
Marca: Cytiva 18115315
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Tricorn columns are designed for high resolution liquid chromatography of biomolecules performed at medium to high pressures
  • Tricorn columns are designed for high resolution liquid chromatography of biomolecules performed at medium to high pressures. They are suitable for use with high performance media, such as Sepharose High Performance, Superdex Prep Grade, and SOURCE.
  • The columns are available with inner diameters of 5 and 10mm, and in lengths from 20 to 200mm. Column lengths of 300 and 600mm are also available for the 10mm diameter columns. The design and dimensions of the columns make them particularly useful for screening and polishing steps in protein purification.
  • Glass tubes coated with plastic film protects individuals in the laboratory from harm if the column is dropped or operated outside its specification.
TRUSTED_SUSTAINABILITY
Specifica
Diametro (sistema metrico) 10 mm
Lunghezza (sistema metrico) 100 mm
Materiale Borosilicate Glass Coated with Protective Plastic Film