Learn More
BRAND™ Borosilicate Glass Conical Ground Joint Stopper
Sfoglia Tutto Brand Prodotti
Click to view available options
Giunto rastremato standard:
NS 12/21
NS 14/23
NS 19/26
NS 29/32
NS 34/35
Dimensione della confezione:
Pezzo
Description
- Borosilicate glass constrcution (Boro 3.3. to DIN 12252)
- Solid or semi-hollow
- Octagonal grip
Specifications
Specifications
| Quantità | 1/Pk. |
| Giunto rastremato standard | NS 14/23 |
| Materiale | Borosilicate Glass |
| Product Type | Ground Joint Stopper |
| Forma | Octagonal Grip (Solid) |