Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with PTFE Plug, Scale
Click to view available options
Capacità (sistema metrico):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
missing translation for 'unitSize'
Pezzo
pezzi 2
Description
- With NS socket and PE stopper
- with PTFE plug
- with screwthread retaining device
- with scale (approx. volume /ml)
- material: borosilicate glass 3.3
Specifications
Specifications
| Capacità (sistema metrico) | 50 mL |
| Materiale | Borosilicate Glass |
| Quantità | 2Pack |
| Stopper Material | Polyethylene |