Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with PTFE Plug, Scale
Codice prodotto. 11846164
Click to view available options
Capacità (sistema metrico):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Dimensione della confezione:
Pezzo
pezzi 2
Codice prodotto. 11846164
Marca: Lenz Laborglasinstrumente™ 04016173
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with PTFE Plug, Scale are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
  • With NS socket and PE stopper
  • with PTFE plug
  • with screwthread retaining device
  • with scale (approx. volume /ml)
  • material: borosilicate glass 3.3
Specifica
Capacità (sistema metrico) 2000 mL
Materiale Borosilicate Glass
Stopper Material Polyethylene