Learn More
Cytiva Borosilicate Glass XK Chromatography Tubes
Codice prodotto. 10516740 Sfoglia Tutto Cytiva Prodotti
Click to view available options
Overall Length (Metric):
20 cm
30 cm
40 cm
60 cm
70 cm
100 cm
400 cm
Da utilizzare con:
16/100 Columns
16/20 Columns
16/40 Columns
16/70 Columns
26/100 Columns
26/20 Columns
26/40 Columns
26/70 Columns
50/100 Columns
50/20 Columns
50/30 Columns
50/60 Columns
Dimensione della confezione:
Pezzo
Codice prodotto. 10516740
Marca: Cytiva 18100085
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Can be used in aqueous solutions and nearly all organic solvents commonly used in liquid chromatography of macromolecules.
  • For use with many organic solvents except chlorinated hydrocarbons, acetone and other ketones, aliphatic esters and phenol
  • Solutions containing more than 10% NaOH, 10% HCl, other strong mineral acids or 5% acetic acid should not be used
TRUSTED_SUSTAINABILITY
Specifica
Da utilizzare con 50/20 Columns
Tipo XK Column Tubes
Overall Length (Metric) 20 cm
Materiale Borosilicate Glass
Min. Operating Temperature (Metric) 4°C
Temperatura operativa max (sistema metrico) 60°C