Learn More
Cytiva Borosilicate Glass XK Chromatography Tubes
Sfoglia Tutto Cytiva Prodotti
Click to view available options
Overall Length (Metric):
20 cm
30 cm
40 cm
60 cm
70 cm
100 cm
400 cm
Da utilizzare con:
16/100 Columns
16/20 Columns
16/40 Columns
16/70 Columns
26/100 Columns
26/20 Columns
26/40 Columns
26/70 Columns
50/100 Columns
50/20 Columns
50/30 Columns
50/60 Columns
Dimensione della confezione:
Pezzo
Descrizione
- For use with many organic solvents except chlorinated hydrocarbons, acetone and other ketones, aliphatic esters and phenol
- Solutions containing more than 10% NaOH, 10% HCl, other strong mineral acids or 5% acetic acid should not be used
Specifica
Specifica
| Da utilizzare con | 26/40 Columns |
| Tipo | XK Column Tubes |
| Diametro (sistema metrico) interno | 26 mm |
| Overall Length (Metric) | 40 cm |
| Materiale | Borosilicate Glass |
| Min. Operating Temperature (Metric) | 4°C |
| Temperatura operativa max (sistema metrico) | 60°C |