Learn More
Corning™ Bottle Top Dispenser (BTD) PTFE Plunger
Codice prodotto. 16802110 Sfoglia Tutto Corning Prodotti
Click to view available options
Volume (sistema metrico):
2.5 mL
5 mL
10 mL
25 mL
50 mL
100 mL
Dimensione della confezione:
Pezzo
Codice prodotto. 16802110
Marca: Corning™ 6861
PTFE plunger for BTD and dispensing reagents
  • Specialized airpurging and closed circuit system ensures no loss of reagent during dispensing, as well as when the device is not in use
  • Comes with four adapters with different diameters for universal use with multiple bottle sizes
  • Telescopic suction tube can be adjusted smoothly to different bottle heights
  • Calibration tool allows easy in-lab calibration with built-in recalibration mechanism to ensure the highest reproducibility during the dispenser’s long life cycle
  • Every dispenser is individually tested and delivered with a certificate of performance
  • Uses:
    • For dispensing organic and inorganic liquids
  • TRUSTED_SUSTAINABILITY
    Specifica
    Volume (sistema metrico) 2.5 mL
    Da utilizzare con (applicazione) Dispensing Organic and Inorganic Liquids
    Include Four adapters