Learn More
Corning™ Bottle Top Dispensers (BTD) Cylinders
Sfoglia Tutto Corning Prodotti
Click to view available options
Volume (sistema metrico):
2.5 mL
5 mL
10 mL
25 mL
50 mL
100 mL
Dimensione della confezione:
Pezzo
Descrizione
- Specialized airpurging and closed circuit system ensures no loss of reagent during dispensing, as well as when the device is not in use
- Comes with four adapters with different diameters for universal use with multiple bottle sizes
- Telescopic suction tube can be adjusted smoothly to different bottle heights
- Calibration tool allows easy in-lab calibration with built-in recalibration mechanism to ensure the highest reproducibility during the dispenser’s long life cycle
- Every dispenser is individually tested and delivered with a certificate of performance
- For dispensing organic and inorganic liquids
Specifica
Specifica
| Volume (sistema metrico) | 25 mL |
| Da utilizzare con (applicazione) | Dispensing Organic and Inorganic Liquids |
| Include | Four adapters |