Learn More
Trajan™ BP1 GC Capillary Columns
Click to view available options
Lunghezza (sistema metrico):
12 m
12 mm
25 m
30 m
50 m
60 m
60 mm
Diametro (sistema metrico) interno:
0.15 mm
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Spessore pellicola:
0.1 μm
0.25 μm
0.5 μm
1 μm
3 μm
5 μm
0.25 mm
Dimensione della confezione:
Pezzo
Descrizione
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
- Classic cross-linked dimethyl polysiloxane technology
- Low bleed
- Non-polar
- Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Specifica
Specifica
| Temperatura max | 320/340°C |
| Diametro (sistema metrico) interno | 0.32 mm |
| Spessore pellicola | 1 μm |
| Lunghezza (sistema metrico) | 25 m |