Learn More
Trajan™ BP1 GC Capillary Columns
Codice prodotto. 12654945
Click to view available options
Lunghezza (sistema metrico):
12 m
12 mm
25 m
30 m
50 m
60 m
60 mm
Diametro (sistema metrico) interno:
0.15 mm
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Spessore pellicola:
0.1 μm
0.25 μm
0.5 μm
1 μm
3 μm
5 μm
0.25 mm
Dimensione della confezione:
Pezzo
Codice prodotto. 12654945
Marca: Trajan™ 054044
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
  • Classic cross-linked dimethyl polysiloxane technology
  • Low bleed
  • Non-polar
  • Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Specifica
Tipo GC Column for Environmental Analysis
Diametro (sistema metrico) interno 0.25 mm
Spessore pellicola 0.25 μm
Lunghezza (sistema metrico) 30 m