Learn More
Novus Biologicals™ BPTF/FALZ Recombinant Protein Antigen
Sfoglia Tutto Bio Techne Prodotti
Click to view available options
Quantità:
100 μL
Dimensione della confezione:
100µL
Descrizione
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human BPTF/FALZ. Source: E.coli Amino Acid Sequence: TTIASTGQTFQITGNPVTMAGKVITKLPLPANSKIVAVNVPATQGGIVQVHQ The BPTF/FALZ Recombinant Protein Antigen is derived from E. coli. The BPTF/FALZ Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifica
Specifica
| Gene ID (Entrez) | 2186 |
| Metodo di purificazione | >80% by SDS-PAGE and Coomassie blue staining |
| Nome comune | BPTF/FALZ Recombinant Protein Antigen |
| Content And Storage | Store at −20°C. Avoid freeze-thaw cycles |
| Formulazione | PBS and 1M Urea, pH 7.4 |
| Da utilizzare con (applicazione) | Blocking/Neutralizing, Control |
| Alias gene | bromodomain and PHD domain transcription factor, Bromodomain and PHD finger-containing transcription factor, bromodomain PHD finger transcription factor, EC 3.6.1, EC 6.2.1.5, FAC1fetal Alz-50 reactive clone 1, FALZnucleosome-remodeling factor subunit BPT |
| Simbolo del gene | BPTF |
| Tipo di etichetta | Unlabeled |
| Product Type | Recombinant Protein Antigen |
| Vedi altri risultati |
For research use only.