Learn More
Brady™ Signs: Emergency Stairs
Click to view available options
Dimensione della confezione:
Pezzo
Descrizione
- ToughStripe™ Floor Signs communicate important safety and warning messages to your employees in areas with foot traffic
- Durable ToughStripe™ Anti-Skid Polyester (B-534) includes a rigid, clear overlaminate and is selectively covered by anti-skid tread
- Highly visible, glossy material resists harsh chemicals and is easy to apply and remove
- Floor signs alert workers of potential hazards, direct traffic, and enforce protective equipment requirements.
Specifica
Specifica
| Certificazione/Conformità | ANSI A1264.2-2006, OSHA 1910.22, ASTM ANSI F1679 |
| Finitura | Gloss |
| Materiale | Polyester |
| Quantità | 1 Ea. |
| Tipo di adesivo | Rubber-based |
| Descrizione | ToughStripe - Pre-printed Floor Signs |
| Diametro (sistema imperiale) | 17 in. |
| Diametro (sistema metrico) | 431.80 mm |
| Da utilizzare con (applicazione) | To alert workers of potential dangers |
| Temperatura max (sistema imperiale) | 130°F |
| Vedi altri risultati |