Learn More
Brady™ Nastro di demarcazione per pavimento ToughStripe™
Click to view available options
Colore:
Arancione
Bianco
Blu
Giallo
Nero
Nero, bianco, White
Nero, giallo, Yellow
Rosso
Rosso, bianco, Red
Verde
Larghezza (sistema imperiale):
2 in.
3 in.
4 in.
Larghezza (sistema metrico):
5.08 cm
7.6 cm
7.62 cm
10.2 cm
Dimensione della confezione:
1 ROLL
1 rotolo
Descrizione
- Visible floor-marking improves safety and organization in almost any industrial environment
- High-gloss surface shines like new paint and withstands forklift traffic without tearing or lifting
- Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
- Can be easily applied by one person working alone
Specifica
Specifica
| Certificazione/Conformità | ASTM ANSI F1679, ANSI A1264.2 2006, OSHA 1910.22 |
| Descrizione | ToughStripe FLOOR TAPES |
| Tipo di adesivo | Gomma permanente, Permanent |
| Rivestimento | None |
| Colore | Nero, bianco, White |
| Finitura | Lucido |
| Formato | Nastro |
| Materiale | Poliestere |
| Printable Sides | Single-side Printable |
| Print Technology | None |
| Show More |