Learn More
Brady™ BradyJet™ Continuous Polyester Labels
Codice prodotto. 16150511
Click to view available options
Dimensions (LxW):
100 ft. x 1.125 in.
100 ft. x 2.25 in.
100 ft. x 4 in.
Dimensione della confezione:
Pezzo
Codice prodotto. 16150511
Marca: Brady™ 150009
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Brady™ BradyJet™ Continuous Polyester Labels are for use with BradyJet™ J2000 and are ideal for a wide range of identification applications.
  • Constructed of indoor-use general purpose polyester
  • Features a specially-formulated topcoat for improved durability and color
  • Specially-formulated topcoat enhances colors and label appearance
  • Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting
  • Topcoat protects print from smudging and harsh conditions
  • Inkjet-receptive labels for use with the BradyJet™ J2000 Color Label Printer
Specifica
Tipo di adesivo Acrylic, Permanent
Rivestimento None
Colore White
Finitura Opaco
Formato Rotolo
Materiale Polyester
Printable Sides Single-side Printable
Print Technology Ink Jet
Product Type Label
Sterilità Non-sterile
Da utilizzare con (applicazione) Pipe and Valve Marking, Safety and Compliance ID, Visual Workplace Labeling, Warehouse Marking, Lockout Tagout Labeling, GHS and Chemical Labels, Arc Flash Labels
Da utilizzare con (apparecchiatura) BradyJet™ J2000 Printer
Etichette per riga 1
Min. Operating Temperature (Metric) -40°C
N. per confezione 1 Label/Bag
Intervallo di temperatura (sistema imperiale) -40°F to +140°F
Temperature Range (Metric) -40°C to +60°C
Spessore (sistema imperiale) 0.0047 in.
Spessore (sistema metrico) 0.119 mm
Dimensions (LxW) 100 ft. x 4 in.
Lunghezza (sistema imperiale) 100 ft.
Lunghezza (sistema metrico) 30.48 m
Larghezza (sistema imperiale) 4 in.
Larghezza (sistema metrico) 101.60 mm
Diametro (sistema imperiale) 1.88 in.
Alternate Part Number J20C-4000-2569