Learn More
Brady™ BradyPrinter i5300 Industrial Label Printer
Click to view available options
Product Type:
Printer
Printer Kit
Tipi di connettore:
Cabled, Ethernet, USB 2.0, USB Host
Cabled, Ethernet, USB 2.0, USB Host, Wi-Fi
Alternate Part Number:
i5300-600-EU-BWS
i5300-600WF-EU-BWS
i5300-C-EU-BWS
i5300-C-EU-WF-BWS
Dimensione della confezione:
1 set
Descrizione
- 300 dpi: prints small fonts with precision
- Requires no calibration and no heat and speed setting adjustments
- 20-second material changeovers, 5.08 mm to 101.6 mm label widths
- Pauses, resumes and stores print jobs on printer in an adjustable queue with overwrite protection
- 178 mm colour touchscreen and built-in auto cutter
- Solid construction with an all-metal frame
- Ships with a licence for the Brady Workstation suite of your preference: Product and Wire Identification, Safety and Facility Identification, or Lab Identification, enabling easy creation of a wide range of labels and signs.
- Intuitive and easy to operate
Specifica
Specifica
| Product Type | Printer |
| Colore | Blue, Gray |
| Tipo di display | Color LCD Touchscreen |
| Materiale | Plastic |
| Print Technology | Direct Thermal, Thermal Transfer |
| Lunghezza (sistema imperiale) | 15.8 in. |
| Larghezza (sistema imperiale) | 11.2 in. |
| Lunghezza (sistema metrico) | 401.32 mm |
| Larghezza (sistema metrico) | 284.48 mm |
| Altezza (sistema imperiale) | 11 in. |
| Vedi altri risultati |