Learn More
Thermo Scientific Chemicals Brain Natriuretic Peptide (1-32), Human
Descrizione
Brain natriuretic peptide is originally isolated from brain, but is mainly produced in myoendocrine cells of the heart ventricles from which it is released into the circulation. It is involved in blood pressure control and cardiovascular homeostasis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Specie | Human |
| Metodo di purificazione | HPLC |
| Content And Storage | -30°C to -10°C |
| Formato | Solid |
| Peso molecolare | 3464.03 |
| Product Type | Brain Natriuretic Peptide (1-32), human |
| Quantità | 1 mg |
| Status giuridico | RUO - research use only |
| Sequenza | BNP (1-32), human |
RUO – Research Use Only
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.