Learn More
BRAND™ BRANDplates cellGrade™ 96-Well, cellGrade Plus-Treated, Flat-Bottom Microplate
Sfoglia Tutto Brand Prodotti
Click to view available options
Colore:
Black, Clear Bottom
Dimensione della confezione:
pezzi 5
Descrizione
- Hydrophilic surface with freely-accessible amino, carboxyl and hydroxyl groups
- Protein-like composition allows for direct cellular attachment
- Sterilized by β-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
- Meet the requirements of Ph. Eur. and the USP 29
- DNA-, DNase-, RNase-free
- Endotoxins are <0.01 EU/mL
- Orange alphanumeric embossing for easy well identification
Specifica
Specifica
| Certificazione/Conformità | ISO 11137, DIN EN ISO 10993 |
| Imballaggio | 5/Pk. |
| Colore | Black, Clear Bottom |
| Product Type | Plus 96-Well Microplate |
| Materiale | Polystyrene |
| No. of Wells | 96 |
| Sterilità | Sterile |
| Trattamento superficiale | cellGrade Plus |
| Forma pozzetto | Flat Bottom (F-well) |
| Da utilizzare con (applicazione) | Cultivation of cells in reduced serum media and fastidious cell lines |
| Vedi altri risultati |