Learn More
Cole-Parmer™ Brass Compression Pipe Adapter
Codice prodotto. 11825453
Click to view available options
Dimensioni dei tubi:
1/2 in.
3/8 in.
Dimensione della confezione:
Pezzo
Codice prodotto. 11825453
Marca: Cole-Parmer™ 3141238
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Offers very good chemical resistance. Ideal when working with any rigid tubing, including tubing made from fluoropolymers, other rigid plastics and metals
  • Very good chemical resistance.
  • Compression fittings work well with rigid tubing such as PTFE or polypropylene.
  • A compression fitting has a nut that compresses rigid tubing against fitting body for high-pressure applications.
  • Compression connector family is ideal when working with rigid tubing.
  • When selecting your fittings, consider your tubing type, material compatibility and temperature and pressure conditions.
Specifica
Autoclavabile Not Autoclavable
Materiale Brass
Min. Operating Temperature (English) -85°F
Min. Operating Temperature (Metric) -65°C
Dimensioni dei tubi 1/2 in.
Pressione massima 400 psi
Max. Operating Temperature (English) 482°F
Max. Operating Temperature (Metric) 250°C
Product Type Compression x NPT(M) Connector