Learn More
Cole-Parmer™ Brass Straight Compression Union
Codice prodotto. 11775918
Click to view available options
Dimensioni dei tubi:
1/16 in.
1/2 in
1/4 in.
1/8 in.
Dimensione della confezione:
Pezzo
Codice prodotto. 11775918
Marca: Cole-Parmer™ 3141250
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Offers very good chemical resistance. Ideal when working with any rigid tubing, including tubing made from fluoropolymers, other rigid plastics and metals
  • Very good chemical resistance.
  • Compression fittings work well with rigid tubing such as PTFE or polypropylene.
  • A compression fitting has a nut that compresses rigid tubing against fitting body for high-pressure applications.
  • Compression connector family is ideal when working with rigid tubing.
  • When selecting your fittings, consider your tubing type, material compatibility and temperature and pressure conditions.
Specifica
Autoclavabile Not Autoclavable
Materiale Brass
Min. Operating Temperature (English) -85°F
Min. Operating Temperature (Metric) -65°C
Dimensioni dei tubi 1/16 in.
Pressione massima 400 psi
Max. Operating Temperature (English) 482°F
Max. Operating Temperature (Metric) 250°C
Product Type Straight Compression Union