Learn More
Aalborg™ Griglia di sostegno in ottone
Click to view available options
Lunghezza (sistema metrico):
65 mm
150 mm
Dimensione della confezione:
Pezzo
Descrizione
- Rib-guided or fluted metering tubes facilitate stable and accurate readings
- Magnifier lens in front shield to enhance reading resolution
- Interchangeability of Flow Tubes and floats
- Ease of installation and exchange of flow tubes
- Glass flow tubes are prevented from turning during the tightening phase of the assembly procedure
- Chemical compatibility
- Simple means of panel mounting
- Frame for P-meter with plug
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | Flowmeter |
| Tipo | Sostegno del telaio |
| Include | Brass plug |
| Lunghezza (sistema metrico) | 65 mm |
| Larghezza (sistema metrico) | 3.17 mm |
| Warranty | 15 mesi |