Learn More
VEE GEE Brix Hydrometer
Codice prodotto. p-7250820
Click to view available options
Intervallo Brix:
-5 to +5°
0 to 12°
0 to 35°
15 to 25°
19 to 31°
29 to 41°
35 to 70°
39 to 51°
5 to 15°
9 to 21°
Dimensione della confezione:
Pezzo
Codice prodotto. 17896613
Marca: VEE GEE 660115
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
The Brix Hydrometer is calibrated to show the percentage of sucrose by weight at 20°C. The Brix scale is based on the following: 1°Brix = 1% sucrose by weight at the specified temperature (20°C).
  • Approx. Length: 330 mm
Specifica
Tipo Brix Hydrometer
Incrementi 0.1°
Lunghezza (sistema metrico) 33 cm
Intervallo Brix 15 to 25°
Temperature of Standardization 20°C