missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Purple

Codice prodotto. p-7024591
Click to view available options
Quantità:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11474097

Brand: Thermo Scientific Chemicals A19530.09

Richiesta di formato massivo o personalizzato

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Edge
This item is not returnable. View return policy

CAS: 115-40-2 | C21H16Br2O5S | 540.22 g/mol

It is used as a pH indicator and to measure serum albumin concentrations. It is used as a pH indicator (5.2-6.8 : yellow to blue-purple) and also used in spectrophotometric determination of serum-albumin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pH indicator and to measure serum albumin concentrations. It is used as a pH indicator (5.2-6.8 : yellow to blue-purple) and also used in spectrophotometric determination of serum-albumin.

Solubility
Insoluble in water. Soluble in ethanol (80 mg/ml), alcohol, methanol (4 mg/ml), methyl cellosolve (300 mg/ml) and dilute alkalies.

Notes
Transition interval: pH 5.2 (yellow) to pH 6.8 (purple)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-40-2
Formula molecolare C21H16Br2O5S
Molecular Weight (g/mol) 540.22
Numero MDL MFCD00011681
InChI Key ABIUHPWEYMSGSR-UHFFFAOYSA-N
PubChem CID 8273
ChEBI CHEBI:86154
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-6-methylphenol
SMILES CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1

Specifications

Materiale o nome chimico Bromocresol Purple
Punto di fusione ∼240°C (decomposition)
Beilstein 359618
Indice di Merck 14,1387
Informazioni di solubilità Insoluble in water. Soluble in ethanol (80mg/ml),alcohol,methanol (4mg/ml),methyl cellosolve (300mg/ml) and dilute alkalies.
Peso formulazione 540.24
Quantità 10 g

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.