missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromodeoxyuridine/BrdU Antibody (BRD494), Novus Biologicals™
Mouse Monoclonal Antibody
322.00€ - 617.00€
Specifica
| Antigene | Bromodeoxyuridine/BrdU |
|---|---|
| Clone | BRD494 |
| Concentrazione | 0.2 mg/mL |
| Diluizione | Flow Cytometry 0.5-1 ug/million cells, Immunohistochemistry 0.5-1 ug/mL, Immunocytochemistry/ Immunofluorescence 0.5-1 ug/mL, Immunohistochemistry-Paraffin 0.5-1.0ug/mL |
| Applicazioni | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantità | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantità | Price | Quantity & Availability | |||||
|
18400862
|
Novus Biologicals
NBP2-32920-0.02mg |
0.02 mg |
322.00€
0.02mg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
|
18410162
|
Novus Biologicals
NBP2-32920-0.1mg |
0.1 mg |
617.00€
0.10mg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Description
Bromodeoxyuridine/BrdU Monoclonal antibody specifically detects Bromodeoxyuridine/BrdU in Nonspecies specific samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Bromodeoxyuridine/BrdU | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 10 mM PBS with 0.05% BSA | |
| Bromodeoxyuridine/BrdU conjugated to KLH | |
| Primary | |
| Store at 4C. |
| BRD494 | |
| Flow Cytometry 0.5-1 ug/million cells, Immunohistochemistry 0.5-1 ug/mL, Immunocytochemistry/ Immunofluorescence 0.5-1 ug/mL, Immunohistochemistry-Paraffin 0.5-1.0ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Non-species specific | |
| Bromodeoxyuridine, BUdr | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title