Learn More
Paragon Scientific™ BS/IP/MSL Viscometer for Transparent Liquids
Click to view available options
Intervallo viscosità:
0.6 to 3 cSt
2 to 10 cSt
20 to 100 cSt
200 to 1000 cSt
6 to 30 cSt
60 to 300 cSt
600 to 3000 cSt
Dimensione della confezione:
Pezzo
Descrizione
- High Quality glass capillary viscometer
- Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifica
Specifica
| Intervallo viscosità | 2 to 10 cSt |
| Calibrated | No |
| Costante approssimata | 0.01 |
| Numeri ASTM | D 445, D 446 |
| Certificazione/Conformità | ASTM D 445, D 446 and ISO 3104, 3105 |
| Size | 2 |
4 week lead-time