missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BTF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA563299
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: TSLRRLAEAL PKQSVDGKAP LATGEDDDDE VPDLV Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene encodes the basic transcription factor 3. This protein forms a stable complex with RNA polymerase IIB and is required for transcriptional initiation. Alternative splicing results in multiple transcript variants encoding different isoforms. This gene has multiple pseudogenes.
Specifica
| BTF3 | |
| Polyclonal | |
| Unconjugated | |
| BTF3 | |
| 1700054E11Rik; basic transcription factor 3; BETA-NAC; BTF3; BTF3a; BTF3b; NACB; NAC-beta; Nascent polypeptide-associated complex subunit beta; nascent-polypeptide-associated complex beta polypeptide; OK/SW-cl.8; RNA polymerase B transcription factor 3; transcription factor BTF3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 689 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P20290 | |
| BTF3 | |
| Recombinant protein corresponding to Human BTF3. Recombinant protein control fragment (Product #RP-104290). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto