Learn More
Tocris Bioscience™ BTT 3033
Click to view available options
Quantità:
10 mg
50 mg
Dimensione della confezione:
10mg
50mg
Descrizione
BTT 3033 is a selective inhibitor of integrin α2β1 (EC50 = 130 nM for α2β1 binding to collagen I). Binds to the α2I domain. Exhibits selectivity for α2β1 over integrins α3β1, α4β1, α5β1 and αv. BTT 3033 inhibits platelet aggregation to collagen I coated capillaries under flow and inhibits binding of α2-expressing CHO cells to collagen I under shear stress conditions. Neurogenic and thromboxane A2-induced human prostate smooth muscle contraction are also inhibited by BTT 3033.
Specifica
Specifica
| Quantità | 10 mg |
| Product Type | BTT 3033 |
| Purezza | >98% |
| Peso molecolare | 465.5 |
| Inibitori | BTT 3033 |
For Research Use Only