Learn More
Aalborg™ Buna N O-ring Aluminum Frame with Plug
Click to view available options
Size:
2-Tube, 150 mm
3-Tube, 150 mm
4-Tube, 150 mm
5-Tube, 150 mm
Include:
Plug
Plug and Manifold
Dimensione della confezione:
Pezzo
Descrizione
- Rib-guided or fluted metering tubes facilitate stable, accurate readings
- Interchangeability of flow tubes and floats
- Manifolding at inlet or outlet
- Ease of installation and exchange of flow tubes
- Chemical compatibility
- Simple means of mounting
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | Gas/Liquid Flowmeter (Manifold-mount) |
| Tipo | Frame |
| Include | Plug and Manifold |
| Size | 3-Tube, 150 mm |