Learn More
Aalborg™ Buna N O-ring Aluminum Frame with Plug
Codice prodotto. p-8008818
Click to view available options
Size:
2-Tube, 150 mm
3-Tube, 150 mm
4-Tube, 150 mm
5-Tube, 150 mm
Include:
Plug
Plug and Manifold
Dimensione della confezione:
Pezzo
Codice prodotto. 11768528
Marca: Aalborg™ P21A2BA0
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Aalborg™ Buna N O-ring Aluminum Frame with Plug is for use with multiple tube flow meters, which retain most of the unique features associated with single tube units.
  • Rib-guided or fluted metering tubes facilitate stable, accurate readings
  • Interchangeability of flow tubes and floats
  • Manifolding at inlet or outlet
  • Ease of installation and exchange of flow tubes
  • Chemical compatibility
  • Simple means of mounting
Specifica
Da utilizzare con (apparecchiatura) Gas/Liquid Flowmeter (Parallel-mount)
Tipo Frame
Include Plug
Size 2-Tube, 150 mm