missing translation for 'onlineSavingsMsg'
Learn More
Learn More
uvex Busingess Blue PU 84282 Low Shoe - Width 11
Sfoglia Tutto Uvex Prodotti
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
49
50
51
52
Dimensione della confezione:
1 paio
Descrizione
Uvex 1 business Shoe 8428.1 S3 SRC
- Safe, smart, stylish
- Fashionably business look with trendy design
- Generous fit with wide steel toe cap
- Penetration-resistant, non-metallic midsole in accordance with latest safety standards, does not restrict flexibility
- Uvex Climazone technology ensures optimum wearer comfort
- Breathable, water-resistant high-tech microvelour upper material
- Textile lining provides optimum breathability
- Made from synthetic materials, suitable for people allergic to chrome
- Shock absorbing, durable mono density polyurethane sole unit
- Removable comfort insole, antistatic with moisture control system and shock absorption in heel and at front of foot
- Soft padded collar and tongue
- Sole unit of free of lacquer wetting inhibitors such as silicones, phthalates, etc.
- Fulfills ESD requirements, resistance to ground Kleiner 35 megohms
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 8428.2
- Standard EN ISO 20345:2011 S3 SRC
- Sizes: 39 to 52
Specifica
Specifica
| Colore | Blue |
| Materiale | Microvelour |
| Size | 49 |
| Materiale suola | Polyurethane (PU) |
| Materiale dello zoccolo | Steel |
| Monouso | Reusable |
| Certifications/Compliance (Europe) | EN ISO 20345:2011 |
| Sesso | Unisex |
| Fit | Standard Width |
| Modello | Uvex 1 |
| Vedi altri risultati |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto