missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BVES Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA580386
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human BVES/POPDC1.
Blood vessel epicardial substance (BVES), also designated Popeye protein 1, is a transmembrane protein that plays a role in cell-cell interactions and adhesion, specifically at tight junctions. BVES is composed of an extracellular amino-terminus, three transmembrane domains and a cytoplasmic carboxyl-terminus. It is expressed in the developing coronary vascular system, specifically in the proepicardium, migrating epithelial epicardium, delaminated vasculogenic mesenchyme and vascular smooth muscle cells, where it functions to direct development in heart, epithelial and muscle cells during embryogenesis. BVES accumulates at points of cell-cell contact, such as filopodia and cell borders, and promotes adhesion prior to the arrival of E-cadherin.
Specifica
| BVES | |
| Polyclonal | |
| Unconjugated | |
| BVES | |
| blood vessel epicardial substance; bves; hBVES; LGMD2X; mBVES; MGC42413; POP 1; POP1; POPDC 1; Popdc1; popdc1 protein; popeye 1; popeye domain containing 1; popeye domain-containing 1; popeye domain-containing protein 1; Popeye protein 1; zgc:86887 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 11149 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q8NE79 | |
| BVES | |
| A synthetic peptide corresponding to the C-terminus of the Human BVES/POPDC1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto