Learn More
c-Myc Antibody, HRP, Novus Biologicals™
Goat Polyclonal Antibody
Marca: Novus Biologicals NB600-341
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
c-Myc Polyclonal antibody specifically detects c-Myc in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry.
Specifica
| Western Blot, ELISA, Immunofluorescence | |
| Goats were immunized with a synthetic peptide representing amino acid residues 410-419 (EQKLISEEDL) of human c-Myc Antibody conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support. | |
| RUO |
| Goat | |
| 0.1 mL |