Learn More
c-MYC Mouse anti-Human, Unconjugated, Clone: 3D9C12, Proteintech
Mouse Monoclonal Antibody
Marca:  Proteintech 67447-1-IG-20UL
Codice prodotto. 16800086
  • 145.00€ / 20µL
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
The c-MYC antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human c-MYC. This antibody recognizes human, mouse, rat antigen. The c-MYC antibody has been validated for the following applications: FC, IHC, WB, ELISA analysis.
Specifica
Specifica
c-MYC
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunoprecipitation, Immunohistochemistry, Flow Cytometry, ELISA
3D9C12
Unconjugated
20 μL
4609
IgG1