Learn More
Thermo Scientific Chemicals C-Myc Peptide
Codice prodotto. 15458369
Click to view available options
Quantità:
1 mg
5 mg
Dimensione della confezione:
1mg
5mg
Codice prodotto. 15458369
Marca: Thermo Scientific Chemicals J66846.M^
Richiesta di formato massivo o personalizzato
For cellular and molecular biology applications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifica
Metodo di purificazione HPLC
Content And Storage -30°C to -10°C
Formato Solid
Peso molecolare 1203.29
Product Type C-Myc Peptide
Quantità 1 mg
Status giuridico RUO - research use only
Sequenza Glu-Gln-Lys-Leu-Ile-Ser-Glu-Glu-Asp-Leu