Learn More
C-Reactive Protein/CRP Antibody (C7), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NB600-1333H
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoassay, Sandwich ELISA.
Specifica
| C-Reactive Protein/CRP | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| CRP | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human CRP. | |
| 0.2 mL | |
| Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 1401 | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay | |
| C7 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoassay, Sandwich ELISA | |
| C-reactive protein, C-reactive protein, pentraxin-related, MGC88244, pentraxin 1, PTX1MGC149895 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human CRP | |
| Store at 4C in the dark. | |
| IgG1 |