missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ C-Reactive Protein/CRP Antibody Pair [HRP]
Sfoglia Tutto Bio Techne Prodotti
Click to view available options
Quantità:
15 Plates
5 Plates
Dimensione della confezione:
pezzi 15
pezzi 5
Descrizione
This solid phase sandwich ELISA utilizes an antibody specific for Mouse C-Reactive Protein/CRP.
Specifica
Specifica
| Intervallo saggio | 34.38 pg/mL |
| Assay Sensitivity | 34.38-2200 pg/mL |
| Saggio | Sandwich ELISA |
| Coniugato | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | Antibody Pairs |
| Specificità | This solid phase sandwich ELISA utilizes an antibody specific for Mouse C-Reactive Protein/CRP. |
| Target Species | Mouse |
| Da utilizzare con (applicazione) | Sandwich ELISA |
| Alias gene | C-reactive protein, C-reactive protein, pentraxin-related, MGC88244, pentraxin 1, PTX1MGC149895 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction