Learn More
C21orf58 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25024-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
| C21orf58 | |
| Polyclonal | |
| Unconjugated | |
| C21orf58 | |
| C21orf58 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54058 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.51 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P58505 | |
| C21orf58 | |
| C21orf58 Fusion Protein Ag21758 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |