Learn More
C3/C3b/C3c Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 21337-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This antibody raised against 1314-1663 aa of human C3 protein recognize C3 alpha chain, C3b alpha' chain, and C3c alpha' chain fragment 2.
Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection.Specifica
| C3/C3b/C3c | |
| Polyclonal | |
| Unconjugated | |
| C3 | |
| ARMD9, ASP, C3, C3b, C3c, Complement C3, complement component 3, CPAMD1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 24232, 718 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P01024, P01026 | |
| C3 | |
| C3/C3b/C3c Fusion Protein Ag15537 | |
| 150 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |